Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultra Elite™ Pressure-Demand Full Facepieces

Catalog No. p-7163907 Shop All MSA Products
Click to view available options
Size:
Medium
Large

Pressure-demand full facepieces feature nosecup and E-Z don head harness.

  • State-of-the-art mask provides comfort and protection
  • Ergonomic design
  • Distortion-free lens allows wide field-of-view
  • 5-point harness allows easy on/off
  • Unique airflow prevents lens misting
  • Face blank has a single-sealing flange, silicone
  • Speech diaphragm makes communication easy to discern

Specifications

Color Black
Product Type Full Facepiece Respirator
For Use With (Equipment) SAR, SCBA
Includes Nosecup, E-Z don head harness
Material Silicone
Size Medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.