Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Ultra-High Performance Fingertight Fittings, PEEK

Catalog No. p-7191290
Click to view available options
Type:
Fingertight Nut
Micro Ferrule

The innovative line of Upchurch Ultra High Performance fittings are desigined to withstand extreme temperature anf pressures. This line of fittings is perfect for use within the increasingly demanding requirements for today's high performance analytical systems. The fittings are a 10/32 fingertight style for use with 1/16 in. and 1/32 in. OD tubing.

TRUSTED_SUSTAINABILITY

Specifications

Type Micro Ferrule
Color Black
Model LiteTouch
Packaging Quantity 10/Pk.
Description 1/32 in.
For Use With 1/32 Tubing
Material PEEK
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.