Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ UNC 0638

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective inhibitor of G9a and GLP histone lysine methyltransferases (IC50 values are < 15 nM and 19 nM for G9a and GLP respectively, and > 10000 nM for a range of other histone methyltransferases). Potently inhibits dimethylation of H3K9 in MCF-7 cells (IC50 = 70 nM). Cell permeable.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 509.73 |
Purity | >99% |
Molecular Weight (g/mol) | 527.754 |
Chemical Name or Material | UNC 0638 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction