Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ UNC 2327
Click to view available options
Quantity:
10 mg
50 mg
Description
UNC 2327 is an allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 230 nM). Noncompetitive with both the peptide substrate and cofactor.
Specifications
Specifications
Quantity | 10 mg |
Product Type | UNC 2327 |
Purity | >99% |
Molecular Weight (g/mol) | 319.38 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction