Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unc119b Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7211245 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5253 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5253 Supplier Proteintech Group Inc Supplier No. 262011AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mouse Unc119b is related closely to Unc119, a homolog of C. elegans unc119 protein which was first discovered in C. elegans on the basis of a spontaneous mutation affecting locomotion, feeding behavior and chemosensation. Mouse Unc119b shares 90% sequence identity with human UNC119b protein. UNC119b is a myristoyl-binding protein that acts as a cargo adapter: specifically binds the myristoyl moiety of a subset of N-terminally myristoylated proteins and is required for their localization. Myristoyl-binding activity of UNC119b is required to localize NPHP3 to the primary cilium.

Specifications

Antigen Unc119b
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene UNC119B
Gene Accession No. A6NIH7, Q8C4B4
Gene Alias AA407659, AI414892, unc 119 homolog B (C. elegans), Unc119b
Gene Symbols UNC119B
Host Species Rabbit
Immunogen Unc119b Fusion Protein Ag24599
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106840, 611530, 84747
Target Species Canine, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.