Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC13B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | UNC13B |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
UNC13B Polyclonal antibody specifically detects UNC13B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
UNC13B | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS (pH 7.2), 40% Glycerol | |
10497 | |
IgG | |
Immunogen affinity purified |
Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
hmunc13, homolog of rat Munc13 (diacylglycerol-binding), MGC133279, MGC133280, MUNC13, Munc13-2, protein unc-13 homolog B, unc-13 homolog B (C. elegans), unc-13-like (C. elegans), UNC13Unc13h2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PPDLVLQKDHFLGPQESFPEENASSPFTQARAHWIRAVTKVRLQLQEIPDD | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title