Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC13D/Munc 13-4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10041385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10041385 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10041385 Supplier Novus Biologicals Supplier No. NB10041385
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 4 publications

UNC13D/Munc 13-4 Polyclonal specifically detects UNC13D/Munc 13-4 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA, Western Blot.

Specifications

Antigen UNC13D/Munc 13-4
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Immunohistochemistry 2.5 ug/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 ug/mL, Peptide ELISA Detection limit 1:128000, Knockdown Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_954712.1
Gene Alias FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans)
Gene Symbols UNC13D
Host Species Goat
Immunogen Peptide with sequence C-ETQKHKKDLHPLFD corresponding to internal region according to NP_954712.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201294
Test Specificity Expected from sequence similarity: Rat, Dog, Pig and Cow.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.