Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC45B Antibody, Novus Biologicals™
SDP

Catalog No. NBP147199 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147199 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147199 Supplier Novus Biologicals Supplier No. NBP147199
Add to Cart
Add to Cart

Goat Polyclonal Antibody

UNC45B Polyclonal specifically detects UNC45B in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen UNC45B
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_775259.1
Gene Alias CMYA4, FLJ38610, MGC119540, MGC119541, protein unc-45 homolog B, SMUNC45, striated muscle UNC45, unc-45 homolog B (C. elegans), UNC-45B, UNC45cardiomyopathy associated 4
Gene Symbols UNC45B
Host Species Goat
Immunogen Peptide with sequence C-RSDKLRQKIFKER corresponding to internal region according to NP_001028748.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146862
Test Specificity This antibody is expected to recognize both reported isoforms (NP_775259.1and NP_001028748.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.