Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC5H1/Unc5a Antibody (Dettwy-1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP197629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197629 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197629 Supplier Novus Biologicals Supplier No. NBP197629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UNC5H1/Unc5a Monoclonal specifically detects UNC5H1/Unc5a in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen UNC5H1/Unc5a
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone Dettwy-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Gene Alias KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5H1FLJ16449
Gene Symbols UNC5A
Host Species Mouse
Immunogen Extracellular domain of human UNC5A (aa 26-306) fused to the Fc portion of human IgG1.
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90249
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.