Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC84B Antibody (3.1E), PE, Novus Biologicals™
SDP

Catalog No. NB308743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308743 Supplier Novus Biologicals Supplier No. NBP259942PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UNC84B Monoclonal antibody specifically detects UNC84B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen UNC84B
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3.1E
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias FRIGG, KIAA0668, MGC133055, MGC133056, nuclear envelope protein, Protein unc-84 homolog B, Rab5-interacting protein, rab5IP, Sad1 and UNC84 domain containing 2, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, SUN domain-containing protein 2, unc-84 homolog B, unc-84 homolog B (C. elegans), UNC84B
Host Species Mouse
Immunogen GST fused to residues 1-121 of human UNC84B
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 25777
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.