Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC93A Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017169 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017169 Supplier Novus Biologicals Supplier No. NBP281858
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC93A Polyclonal specifically detects UNC93A in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen UNC93A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, ELISA
Formulation PBS
Gene Alias dJ366N23.1, dJ366N23.1 (putative C. elegans UNC-93 (protein 1, C46F11.1) LIKE), dJ366N23.2, dJ366N23.2 (putative C. elegans UNC-93 (protein 1, C46F11.1) C-terminal LIKE), unc93 homolog A, unc-93 homolog A (C. elegans)
Gene Symbols UNC93A
Host Species Rabbit
Immunogen UNC93A antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human UNC93A. The immunogen is located within the last 50 amino acids of UNC93A. Amino Acid Squence: CVESKNPIRPHAPGQV
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54346
Test Specificity Two isoforms of Unc93a are known to exist. This antibody will not cross-react with Unc93b.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.