Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC93A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB342112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342112 Supplier Novus Biologicals Supplier No. NBP281858PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC93A Polyclonal antibody specifically detects UNC93A in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen UNC93A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias dJ366N23.1, dJ366N23.1 (putative C. elegans UNC-93 (protein 1, C46F11.1) LIKE), dJ366N23.2, dJ366N23.2 (putative C. elegans UNC-93 (protein 1, C46F11.1) C-terminal LIKE), unc93 homolog A, unc-93 homolog A (C. elegans)
Host Species Rabbit
Immunogen UNC93A antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human UNC93A. The immunogen is located within the last 50 amino acids of UNC93A. Amino Acid Squence: CVESKNPIRPHAPGQV
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54346
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.