Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC93B Antibody, Novus Biologicals™
SDP

Catalog No. NBP224743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224743 0.1 mg
NBP224743SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224743 Supplier Novus Biologicals Supplier No. NBP224743
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

UNC93B Polyclonal specifically detects UNC93B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UNC93B
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26067373)
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Alias hUNC93B1, MGC126617, unc93 (C. elegans) homolog B1, unc93 homolog B, unc93 homolog B1, unc-93 homolog B1 (C. elegans), unc-93 related protein, UNC93B, Unc-93B1, UNC93protein unc-93 homolog B1
Gene Symbols UNC93B1
Host Species Rabbit
Immunogen A portion of amino acid 500-550 of mouse UNC93B protein was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81622
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse, 77% homologous in opossum and 72% homologous in xenopus.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.