Learn More
Description
Specifications
Specifications
| Antigen | UNC93B |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 3-5 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 26067373) |
| Formulation | 0.1 ml PBS with 0.05% Sodium Azide |
| Gene Alias | hUNC93B1, MGC126617, unc93 (C. elegans) homolog B1, unc93 homolog B, unc93 homolog B1, unc-93 homolog B1 (C. elegans), unc-93 related protein, UNC93B, Unc-93B1, UNC93protein unc-93 homolog B1 |
| Gene Symbols | UNC93B1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
