Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNCX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
| Antigen | UNCX |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19131420
![]() |
Novus Biologicals
NBP19131420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP191314
![]() |
Novus Biologicals
NBP191314 |
100 μL |
Each for $499.50
|
|
|||||
Description
UNCX Polyclonal specifically detects UNCX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| UNCX | |
| Polyclonal | |
| Rabbit | |
| NP_001073930 | |
| 340260 | |
| Synthetic peptide directed towards the C terminal of human UNCX. Peptide sequence AAATEGGGGDCADAGTAGPAPPPPAPSPRPGPRPPSPAEEPATCGVPEPG. | |
| Primary |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| homeobox protein unc-4 homolog, homeobox protein Uncx4.1, UNC homeobox | |
| UNCX | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title