Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Undecanoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
112-37-8
C11H22O2
186.29
MFCD00002730
ZDPHROOEEOARMN-UHFFFAOYSA-N
hendecanoic acid,undecylic acid,n-undecanoic acid,n-undecoic acid,n-undecylic acid,undecoic acid,undecanoate,1-decanecarboxylic acid,undekansaeure,unii-138on3iiqg
8180
CHEBI:32368
undecanoic acid
CCCCCCCCCCC(=O)O
112-37-8
C11H22O2
186.29
MFCD00002730
ZDPHROOEEOARMN-UHFFFAOYSA-N
hendecanoic acid,undecylic acid,n-undecanoic acid,n-undecoic acid,n-undecylic acid,undecoic acid,undecanoate,1-decanecarboxylic acid,undekansaeure,unii-138on3iiqg
8180
CHEBI:32368
undecanoic acid
CCCCCCCCCCC(=O)O
Packaging Glass bottle
Quantity 100g
Boiling Point 284.0°C
Flash Point >110°C
Melting Point 28.5°C
Infrared Spectrum Authentic
Color White to Yellow
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)9CO2H
Beilstein 02, 358
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,cresols and 1-butanol
Formula Weight 186.29
Physical Form Low Melting Solid
Percent Purity 99%
Chemical Name or Material Undecanoic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-964-2

RTECSNumber : YQ2275000

TSCA : TSCA