Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNG Antibody (OTI1A11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274785A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274785A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274785A700 Supplier Novus Biologicals Supplier No. NBP274785AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UNG Monoclonal specifically detects UNG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UNG
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A11
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DGUEC 3.2.2, HIGM4EC 3.2.2.27, UDGDKFZp781L1143, UNG15, UNG1uracil-DNA glycosylase 1, uracil-DNA glycosylase 2, UNG2, uracil-DNA glycosylase
Gene Symbols UNG
Host Species Mouse
Immunogen Full length recombinant protein of human UNG (NP_550433) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Base Excision Repair, Cancer, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7374.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.