Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ UniSafe™ Vented Wash Bottle

Shop All United Scientific Supplies, Inc Products


Click to view available options
Chemical Label:
Acetone
Acetonitrile
Dichloromethane
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methanol
Methyl Ethyl Ketone
Sodium Hypochlorite
Toluene
Water
Description
Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Each bottle is compliant with GHS and OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200. Intended for use only with LDPE and PP compatible solvents.
- Industrial quality low density polyethylene (LDPE) construction with long lasting strength. GHS compliant
- Preprinted bottles include: chemical name and formula, NFPA diamond, health hazards, and CAS number
- Polypropylene stem closure and draw tube
- Safety vented and color coded for quick solvent recognition
- 4-color printing, 500 mL (16 oz.) capacity
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 16 oz. |
| Color | Blue |
| For Use With (Application) | LDPE and PP-compatible Solvents |
| Material | LDPE |
| Packaging | 6/Pk. |
| Sterility | Non-sterile |
| Certifications/Compliance | GHS and OSHA |
| Closure Included | Yes |
| Disposable | Reusable |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction