Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Shop All Simport Scientific Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Description
- In Quickload™ stacks
- 35° Anterior writing area
- One-piece construction with hinge-lock design prevent early separation of base and lid
- Snap latch and hinge-lock design - cassettes will always relock securely
- Allows for one-hand operation
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 25 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
| Quantity | 1000/Pk. |
| Color | White |
| Material | Acetal |