Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Photoelectric Effect Apparatus

Shop All United Scientific Supplies, Inc Products


Click to view available options
Grade:
5+
Description
- Set of equipment for studying photo-emission of a mixed alkali photocathode.
Specifications
Specifications
Grade | 5+ |
Format | Kit |
Product Type | Photoelectric Effect Apparatus |
Languages | English |
For Use With (Application) | For studying Photo-emission of a mixed alkali photocathode, with illumination and bias control |
Subject | Physics |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction