Learn More
United Scientific™ Polypropylene Laboratory Scoops
Designed for transferring powdered or granular materials.
Supplier: United Scientific™ 81258
Description
- Available in variety of sizes and as complete set of 7 scoops
- Autoclavable material, do not rust or corrode
- Capacities moulded into top of each scoop for easy reference
Specifications
6 Pk. | |
Autoclavable | |
500 mL | |
Polypropylene |
Scoop | |
16.9 oz. | |
Laboratory Scoop | |
318 mm |