Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Universal Press-Tight™ Connectors: Universal Y, Angled
SDP

Catalog No. 06611159 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Includes:
Deactivation
Siltek Deactivated
Undeactivated
Quantity:
1 Ea.
3 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
06-611-159 Deactivation 1 Ea.
06-717-753 Undeactivated 3 Pk.
06-611-158 Undeactivated 1 Ea.
06-611-162 Siltek Deactivated 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-611-159 Supplier Restek™ Supplier No. 20403261
Add to Cart
Add to Cart

Connect analytical columns, guard columns, transfer lines or detectors

  • Split column flow to two detectors
  • Inlet and outlet ends match column curvature to alleviate connection strain
  • Durable Leak-Tight Seal
  • High Temperature Polyamide

Specifications

Type Universal Angled Y Press-Tight Connector
Includes Deactivation
Quantity 1 Ea.
Material Glass
Color Clear
For Use With (Application) Perform Dual-Column Confirmation Analysis with a Single Injection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.