Learn More
Description
- Split column flow to two detectors
- Inlet and outlet ends match column curvature to alleviate connection strain
- Durable Leak-Tight Seal
- High Temperature Polyamide
Specifications
Specifications
| Type | Universal Angled Y Press-Tight Connector |
| Includes | Siltek Deactivated |
| Quantity | 1 Ea. |
| Material | Glass |
| Color | Clear |
| For Use With (Application) | Perform Dual-Column Confirmation Analysis with a Single Injection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
