Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unleaded Gasoline Composite Standard, Restek
SDP

Catalog No. 06728657 Shop All Restek Corporation Products
Click to view available options
Quantity:
1 mL
Concentration or Composition (by Analyte or Components):
50,000 μg/mL

Catalog No. 06-728-657

Supplier: Restek Corporation 30205

Only null left
Add to Cart
Add to Cart

Specifications

Packaging Ampule
Solvent or Matrix Methanol-P and T
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 50,000 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.