Learn More
OpGen Unyvero A50 Analyzer
Catalog No. 23015306
Click to view available options
:
Type: Analyzer
This item is not returnable. View return policy
This item is not returnable. View return policy
This is a rapid sample-to-answer high multiplexed molecular diagnostic system for simultaneous detection of a broad range of pathogens and AMR genes directly from native samples.
  • Benchtop molecular diagnostic system, modular and scalable: Up to 4 Analyzers and 2 Lysators can be connected to 1 Cockpit
  • Rapid, sample-to-answer results achieved with highly multiplexed automated closed-system PCR technology
  • Random access capability allows for on-demand testing; 2 samples at a time per Analyzer
  • Simple to use, fully barcoded cartridge-based technology enables testing during all laboratory shifts
  • Only 2 minutes of hands-on time to set up a run
  • Easy to train laboratory personnel to operate the Unyvero System
  • Can be integrated into LIS (unidirectional)
  • Simultaneously detects a broad range of clinically relevant pathogens and antibiotic resistance markers (AMR) in a single rapid test in just a few hours
  • Direct from native sample testing provides faster results; days earlier than culture because no growth is required for detection with Unyvero molecular technology
  • Faster and more sensitive than culture and traditional microbiological methods because results are not impacted by prior antibiotic use
  • Detects more potential pathogens missed by culture, including multi-pathogen infections as well as pathogens not covered by empiric therapy
  • Clear, qualitative results based on defined cut-offs
  • Moderate CLIA complexity
Specifications
Product Type Unyvero Molecular System
Unyvero Analyzer (60001) is a required component of Unyvero System which consists of a minimum of 1 Unyvero Analyzer and 1 Unyvero Lysator (60002) and 1 Unyvero Cockpit (60003)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.