Learn More
OpGen Unyvero C8 Cockpit
Click to view available options
:
Type: Cockpit
Description
- Benchtop molecular diagnostic system, modular and scalable: Up to 4 Analyzers and 2 Lysators can be connected to 1 Cockpit
- Intuitive central user interface with step-by-step on-screen tutorial; connects Unyvero Analyzer and Lysator
- Manages the testing process and stores results
- Easy to navigate software interface supports ease of use and efficient training of laboratory personnel to operate the Unyvero System during all laboratory shifts
- Can be integrated into LIS (unidirectional)
Specifications
Specifications
Product Type | Unyvero Molecular System |
Unyvero Cockpit (60003) is a required component of Unyvero System which consists of a minimum of 1 Unyvero Analyzer (60001) and 1 Unyvero Lysator (60002) and 1 Unyvero Cockpit (60003)