Learn More
OpGen Unyvero L4 Lysator
Click to view available options
:
Type: Lysator
Description
- Benchtop molecular diagnostic system, modular and scalable: Up to 4 Analyzers and 2 Lysators can be connected to 1 Cockpit
- Lysator performs automated preparation of native samples (liquefaction, cell lysis, homogenization) in a closed-system manner; no open amplicon
- 4 samples at a time per Lysator
- Simple to use technology enables testing during all laboratory shifts
- Easy to train laboratory personnel to operate the Unyvero System
Specifications
Specifications
Product Type | Unyvero Molecular System |
Unyvero Lysator (60002) is a required component of Unyvero System which consists of a minimum of 1 Unyvero Analyzer (60001) and 1 Unyvero Lysator (60002) and 1 Unyvero Cockpit (60003)