Learn More
OpGen Unyvero M1 Master Mix Tube Kit
For use with FDA-cleared Unyvero LRT BAL Cartridge Kit.
Supplier: OpGen 10020
Alert:
Storage: -20°C or below.
Description
- 12 tests per kit; storage: -20°C or below
- Ready-to-use, no prep or mixing required
Specifications
Primer-probe | |
End-point PCR | |
Pathogen Detection |
Frozen | |
Standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unyvero M1 Master Mix Tube Kit (10020), storage: -20°C or below, runs with Unyvero LRT BAL Cartridge Kit (10091) and it is performed on Unyvero System which consists of a minimum of 1 Unyvero Analyzer (60001) and 1 Unyvero Lysator (60002) and 1 Unyvero Cockpit (60003)