Learn More
OpGen Unyvero UTI Cartridge Set (RUO)
Simultaneously identifies 40 pathogens including C. auris and antibiotic resistance markers associated with urinary tract infections.
Supplier: OpGen 10079
Alert:
Unyvero UTI Cartridge Set *RUO* is not cleared with the U.S. Food and Drug Administration (FDA). The Unyvero UTI Cartridge Set *RUO* is for Research Use only. Not for use in diagnostic procedures. Storage: 15-25°C.
Description
- 12 tests per kit; storage: 15-25°C
- Unyvero UTI Cartridge Set *RUO* is not cleared with the U.S. Food and Drug Administration (FDA). The Unyvero UTI Cartridge Set *RUO* is for Research Use only. Not for use in diagnostic procedures.
- Rapid, sample-to-answer results achieved with highly multiplexed automated closed-system PCR technology
- Panel consists of 40 targets (25 pathogens and 15 resistance genes)
- Random access capability allows for on-demand testing
- Simple to use, fully barcoded cartridge-based technology enables testing during all laboratory shifts
- Only 2 minutes of hands-on time to set up a run
- Easy to train laboratory personnel to operate the Unyvero System
- Simultaneously detects a broad range of urinary tract infection pathogens and their associated antibiotic resistance markers (AMR) in a single rapid test in just a few hours
- Direct from urine sample testing provides faster results; days earlier than culture because no growth is required for detection with Unyvero molecular technology
- Pathogens (25): Acinetobacter baumannii complex, Bacteroides spp. / Prevotella spp., Citrobacter freundii / koseri, Coagulase negative staphylococcus, Corynebacterium urealyticum, Enterococcus spp., Enterococcus faecalis, Enterococcus faecium, Enterobacteriaceae/Enterobacterales, Enterobacter cloacae complex, Escherichia coli, Klebsiella aerogenes (Enterobacter aerogenes), Klebsiella oxytoca, Klebsiella pneumoniae, Klebsiella variicola, Proteus spp., Providencia spp., Pseudomonas aeruginosa, Staphylococcus aureus, Staphylococcus saprophyticus, Streptococcus agalactiae FUNGI:, Candida spp., Candida albicans, Candida auris, Candida glabrata
- Resistance genes (15): Oxacillin: mecA; Glycopeptides: vanA, vanB; 3rd generation Cephalosporins: ctx-M; Carbapenem: imp, kpc, ndm, oxa-23, oxa-24, oxa-48, vim; Polypeptides / Polymyxins: mcr-1; Fluoroquinolones: qnrB, qnrS; Sulfonamide: sul1
Specifications
Primer-probe | |
End-point PCR | |
Pathogen Detection |
Dried Down | |
Standard |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unyvero UTI Cartridge Set *RUO* (10079), storage: 15-25°C, runs with Unyvero M1 Master Mix Tube Set *RUO* (10011) and it is performed on Unyvero System which consists of a minimum of 1 Unyvero Analyzer (60001) and 1 Unyvero Lysator (60002) and 1 Unyvero Cockpit (60003).