Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

uPAR Antibody (014), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB333182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333182 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333182 Supplier Novus Biologicals Supplier No. NBP290454JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for ELISA

Specifications

Antigen uPAR
Applications ELISA
Classification Monoclonal
Clone 014
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse uPAR (Accession#: NP_035243.1, Met1-Thr297)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5329
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.