Learn More
MilliporeSigma™ Supelco™ Upchurch PEEK LiteTouch™ Fitting, PEEK
Unique design prevents twisting of PEEK tubing.
$46.25 - $64.00
Specifications
Color Natural
Packaging Quantity 10/Pk.
Material PEEK
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
11-100-5094
View Documents
MilliporeSigma™ Supelco™
57653
Double-winged nut
Each for $46.25
Only null left
Add to Cart
11-100-5093
View Documents
MilliporeSigma™ Supelco™
57651
Nut
Each for $64.00
Only null left
Add to Cart
Description
Description
The fitting consists of a special PEEK ferrule, a stainless steel ring, and a PEEK or stainless steel nut for 1/16 in. tubing. The fitting will not leak at pressures to 5000 psi/340 bar with the PEEK nut, 2500 psi/175 bar with the stainless steel nut finger-tightened or 7000 psi/480 bar with wrench tightening. After wrench-tightening the stainless steel nut, subsequent finger-tightening will hold to 5000 psi. Compatible with all manufacturers' stainless steel nuts. Order nuts and ferrules separately.
Specifications
Specifications
Natural
10/Pk.
PEEK
Videos
SDS
Documents
Documents
Product Certifications
LiteTouch is a registered trademark of IDEX Health & Science, LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.