Learn More
Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID
Description
Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.
- Precut 316 stainless steel tubing:
- Ends are machine cut and polished
- Edges are deburred inside and outside
- Tubing is passivated
- Inside is flushed with reagent-grade isopropanol
- Color-coded banding for easy identification of the inner diameter
- All stainless-steel tubing longer than 1m is coiled
Specifications
Specifications
| Type | Tubing |
| Diameter (English) Inner | 1/16 in. |
| Diameter (English) Outer | 0.040 in. |
| Diameter (Metric) Inner | 1.0 mm |
| Overall Length (English) | 6 in. |
| Overall Length (Metric) | 10 cm |
| Material | Stainless Steel (316) |
| Maximum Pressure | 8100 psi (558 bar) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.