Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UPF3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | UPF3A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15726220
![]() |
Novus Biologicals
NBP15726220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157262
![]() |
Novus Biologicals
NBP157262 |
100 μL |
Each for $487.50
|
|
|||||
Description
UPF3A Polyclonal specifically detects UPF3A in Human samples. It is validated for Western Blot.Specifications
UPF3A | |
Polyclonal | |
Rabbit | |
Human | |
HUPF3A, Nonsense mRNA reducing factor 3A, regulator of nonsense transcripts 3A, RENT3AUp-frameshift suppressor 3 homolog A, UPF3 regulator of nonsense transcripts homolog A (yeast), UPF3hUpf3 | |
UPF3A | |
IgG | |
55 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9H1J1 | |
65110 | |
Synthetic peptides corresponding to UPF3A(UPF3 regulator of nonsense transcripts homolog A (yeast)) The peptide sequence was selected from the middle region of UPF3A (NP_075387). Peptide sequence QRYHVDDGRRHRAHHEPERLSRRSEDEQRWGKGPGQDRGKKGSQDSGAPG. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title