Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
UQCC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | UQCC1 | 
|---|---|
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Form | Liquid | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        50-157-0407
                    
                    
                        
                    
                    
                        
                         
             | 
            
                
                    
                        Bethyl Laboratories, Inc
                         A305430AT  | 
                
            
            
            
            
            
                
                    20 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $151.31
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.Specifications
| UQCC1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Basic FGF-repressed Zic-binding protein, bFGF-repressed Zic-binding protein, BFZB, C20orf44, CBP3, chromosome 20 open reading frame 44, cytochrome B protein synthesis 3 homolog, cytochrome B protein synthesis 3 homolog (S. cerevisiae), FLJ10850, ubiquinol-cytochrome c reductase complex chaperone, ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog, UQCC | |
| Between 249 and 299 | |
| Primary | |
| 4° C | |
| UQCC1 | 
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9CWU6, Q9NVA1 | |
| 55245, 56046 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title