Learn More
Invitrogen™ UQCRFS1 Polyclonal Antibody

Description
Predicted to react with chicken and mouse based on sequence homology.
Specifications
Specifications
| Antigen | UQCRFS1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Uqcrfs1 |
| Gene Accession No. | P47985 |
| Gene Alias | 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1 |
| Gene Symbols | Uqcrfs1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.