Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uranine (Concentrated, Water Soluble/Laboratory), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
| Melting Point | 320°C |
| Identification | Pass Test |
| Formula Weight | 376.28 |
| Grade | Laboratory |
| Packaging | Poly Bottle |
| Color | Orange |
| Quantity | 500 g |
| Physical Form | Powder |
| Chemical Name or Material | Uranine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction