Learn More
Urea, 98+%
Description
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as a protein denaturantUrea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.
Solubility
Soluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.
Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 130°C to 135°C |
| Density | 1.335 |
| pH | 9.2 to 9.5 |
| Odor | Ammonia-like |
| Quantity | 10,000 g |
| Beilstein | 635724 |
| Merck Index | 14,9867 |
| Solubility Information | Soluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether. |
| Formula Weight | 60.06 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| EINECSNumber | 200-315-5 |
| RTECSNumber | YR6250000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.