Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urethane, Thermo Scientific™

Catalog No. AC140762500
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-79-6
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.094
InChI Key JOYRKODLDBILNP-UHFFFAOYSA-N
Synonym urethane, urethan, ethylurethane, carbamic acid ethyl ester, ethyl urethane, carbamic acid, ethyl ester, ethylcarbamate, pracarbamine, leucethane, pracarbamin
PubChem CID 5641
ChEBI CHEBI:17967
IUPAC Name ethyl carbamate
SMILES CCOC(=O)N

Specifications

Assay Percent Range 99%
Linear Formula H2NCO2C2H5
Quantity 250 g
Beilstein 03, 22
Fieser 01,1270
Merck Index 15, 10059
Formula Weight 89.09
Chemical Name or Material Urethane, 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.