Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Uric Acid Liquid Reagents

Catalog No. p-319070
Click to view available options
Quantity:
1 x 120 mL
1 x 500 mL
1 x 1 L
Form:
Liquid
Formulation:
4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
4-Aminoantipyrine >0.3mM, TBHBA >1mM, Uricase 150U/L, Peroxidase >2500U/L, Buffer, non-reactive Stabilizers and fillers
pH:
7.5
8.1
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent requires Uric Acid Standard (23-666-424)/Chemistry Calibrator (23-666-189)

Method: Uricase/trinder, endpoint
Storage Temperature: 2–8°C
Reconstituted Stability: Ready-to-use liquid
Linearity: 20mg/dL
Measurement Wavelength: 520nm
Reaction Temperature: 37°C
Expected Values: 2.5–7.7mg/dL

Specifications

Certifications/Compliance CE Marked
Type Uric Acid Reagent Set
Formulation 4-AAP >0.2mM, HDCBS 2mM, Uricase (microbial) >150U/L, Peroxidase (horseradish) >2500U/L, pH 7.5 ± 0.1 Buffer, non-reactive Stabilizers
For Use With (Application) Quantitative determination of Uric Acid in Serum, in-vitro diagnostic use only
pH 7.5
Quantity 1 x 500 mL
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.