Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uricase Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB100202201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100202201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100202201 Supplier Novus Biologicals Supplier No. NB10020220.1MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Uricase Polyclonal specifically detects Uricase in Bacteria samples. It is validated for ELISA, Immunoprecipitation, Western Blot.

Specifications

Antigen Uricase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias UOXP, urate oxidase, pseudogene, URICASE
Gene Symbols UOX
Host Species Goat
Immunogen Full length native protein: Uricase (Bacillus Genus)
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391051
Test Specificity Recognizes uricase.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.