Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB600469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600469 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600469 Supplier Novus Biologicals Supplier No. NB600469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

URM1 Polyclonal specifically detects URM1 in Yeast samples. It is validated for Western Blot, ELISA.

Specifications

Antigen URM1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:2000-1:10000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P40554
Gene Alias C9orf74, chromosome 9 open reading frame 74, MGC2668, ubiquitin related modifier 1, ubiquitin related modifier 1 homolog, ubiquitin related modifier 1 homolog (S. cerevisiae), ubiquitin-related modifier 1 homolog
Gene Symbols URM1
Host Species Rabbit
Immunogen This purified URM1 Antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast URM1 protein. (Uniprot: P40554)
Molecular Weight of Antigen 11.03 kDa
Purification Method Multi-step
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 81605
Test Specificity Urm 1
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Yeast
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.