Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urocortin Antibody (2D7), Novus Biologicals™
SDP

Catalog No. NB019778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019778 0.025 mg
NBP262804 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019778 Supplier Novus Biologicals Supplier No. NBP2628040.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Urocortin Monoclonal specifically detects Urocortin in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Urocortin
Applications Western Blot, In Situ Hybridization (ISH)
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoassay
Formulation PBS containing 0.1% BSA with No Preservative
Gene Alias MGC129974, MGC129975, urocortin, urocortin, preproprotein, UROCUI
Gene Symbols UCN
Host Species Mouse
Immunogen IMAC-purified SUMO-eXact-preproUCN2
Purification Method Protein G purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 7349
Target Species Human
Content And Storage Store at 4degreesC.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.