Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Urocortin Antibody (2D7), Novus Biologicals™
SDP

Catalog No. NBP262804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262804 100 μg
NB019778 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP262804 Supplier Novus Biologicals Supplier No. NBP262804
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Urocortin Monoclonal specifically detects Urocortin in Human samples. It is validated for Western Blot, Immunoassay.

Specifications

Antigen Urocortin
Applications In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoassay
Formulation PBS containing 0.1% BSA with 0.02% Sodium Azide
Gene Alias MGC129974, MGC129975, urocortin, urocortin, preproprotein, UROCUI
Gene Symbols UCN
Host Species Mouse
Immunogen IMAC-purified SUMO-eXact-preproUCN2
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7349
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.