Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Urolithin A
GREENER_CHOICE

Catalog No. p-7161691
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC)

Urolithin A is a PI 3-K/Akt/mTOR signaling pathway inhibitor; blocks phosphorylation of Akt and p70S6K in pancreatic ductal adenocarcinoma (PDAC). Inhibits proliferation and migration and promotes apoptosis of PDAC cell lines in vitro (IC50 values are 10.23 - 62.28 μM). Suppresses tumor growth and improves survival in mice bearing pancreatic tumor xenografts. Also blocks amyloid-β fibrillation in vitro.

Specifications

Chemical Name or Material 3,8-Dihydroxy-6H-dibenzo[b,d]pyran-6-one
CAS 1143-70-0
Quantity 10 mg
Molecular Formula C13H8O4
Formula Weight 228.2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.