Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Antibody (UPK1A/2923), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB390305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390305 Supplier Novus Biologicals Supplier No. NBP308826IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Uroplakin Ia Monoclonal antibody specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen Uroplakin Ia
Applications ELISA, Protein Array
Classification Monoclonal
Clone UPK1A/2923
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Host Species Mouse
Immunogen Recombinant fragment (around aa 114-173) of human Uroplakin Ia (UPK1A) protein (exact sequence is proprietary) (Uniprot: O00322)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11045
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.