Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Antibody (UPK1A/8704R), Novus Biologicals™
SDP

Catalog No. NB235732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB235732 100 μg
NB235367 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB235732 Supplier Novus Biologicals Supplier No. NBP320317100UG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Uroplakin Ia Monoclonal antibody specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Uroplakin Ia
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UPK1A/8704R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Host Species Rabbit
Immunogen Recombinant fragment (around aa 100-200) of human Uroplakin Ia protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11045
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.