Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ia Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB028346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB028346 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB028346 Supplier Novus Biologicals Supplier No. NBP29322920UL

Rabbit Polyclonal Antibody

Uroplakin Ia Polyclonal antibody specifically detects Uroplakin Ia in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Uroplakin Ia
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-230 of human UPK1A (NP_008931.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11045
Target Species Mouse, Rat
Content And Storage Store at -20°:C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.