Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uroplakin Ib Antibody (UPK1B/3102) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB012890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB012890 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB012890 Supplier Novus Biologicals Supplier No. NBP279934
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Uroplakin Ib Monoclonal specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen Uroplakin Ib
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone UPK1B/3102
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with No Preservative
Gene Alias tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b
Gene Symbols UPK1B
Host Species Mouse
Immunogen Recombinant fragment (around aa 109-229) of human Uroplakin Ib (UPK1B) protein (exact sequence is proprietary) (Uniprot: O75841)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.