Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius USB Data Cable
GREENER_CHOICE

Catalog No. p-7229311 Shop All Sartorius Products
Click to view available options
Connection Type:
USB-C to USB-A
USB-C to USB-B

USB Data Cable for Entris™ Lab Balance.

  • To connect Sartorius laboratory balances of the product family Entris™ to a printer or a second display via USB interface
  • Color: Black
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Weights and Weight Sets
Connection Type USB-C to USB-B
Length (English) 5 ft.
Length (Metric) 1.5 m
Type Data Cable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.