Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Use1/UBE2Z Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB097429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097429 Supplier Novus Biologicals Supplier No. NBP297553AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use1/UBE2Z Polyclonal specifically detects Use1/UBE2Z in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Use1/UBE2Z
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias MDS032, P31, protein p31, putative MAPK activating protein PM26, Putative MAPK-activating protein PM26, Q-SNARE, SLT1, SNARE-like tail-anchored protein 1 homolog, unconventional SNARE in the ER 1 homolog (S. cerevisiae), USE1L, USE1-like protein, vesicle transport protein USE1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Use1/UBE2Z fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55850
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.